Order Entry
Poland
ContactUsLinkComponent
 
Anti-ErbB 2 (pTyr1248) Rabbit Recombinant Antibody [clone: R03-6A8] ()
Catalog # ABNORAB01673.100UL
Supplier:  Abnova
Anti-ErbB 2 (pTyr1248) Rabbit Recombinant Antibody [clone: R03-6A8] ()
Catalog # ABNORAB01673.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    ErbB 2
  • Clonality:
    Recombinant
  • Clone:
    R03-6A8
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Modification:
    pTyr1248
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against synthetic phosphopeptide corresponding to residues surrounding Tyr1248 of human ErbB 2
  • Pk:
    100 µl

Specifications

About this item