Anti-TrkB (pTyr516) Rabbit Recombinant Antibody [clone: R04-3D9] ()
Catalog # ABNORAB01675.100UL
Supplier: Abnova
Specifications
- Antigen symbol:TrkB
- Clonality:Recombinant
- Clone:R04-3D9
- Host:Rabbit
- Reactivity:Human
- Modification:pTyr516
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic phosphopeptide corresponding to residues surrounding Tyr516 of human TrkB
- Pk:100 µl