Anti-Integrin alpha V Rabbit Recombinant Antibody [clone: R02-6G3]
ABNORAB01726.100UL
: Abnova
- Antigen symbol:Integrin alpha V
- Clonality:Recombinant
- Clone:R02-6G3
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human Integrin alpha V
- Pk:100 µl