Anti-PD-L2 Rabbit Recombinant Antibody [clone: R02-2A1]
Catalog # ABNORAB01742.100UL
Supplier: Abnova
Specifications
- Antigen symbol:PD-L2
- Clonality:Recombinant
- Clone:R02-2A1
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human PD-L2
- Pk:100 µl