Order Entry
Poland
ContactUsLinkComponent
Anti-NTHL1 Rabbit Recombinant Antibody [clone: R08-8E9]
Anti-NTHL1 Rabbit Recombinant Antibody [clone: R08-8E9]
Catalog # ABNORAB02048.100UL
Supplier:  Abnova
Anti-NTHL1 Rabbit Recombinant Antibody [clone: R08-8E9]
Catalog # ABNORAB02048.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    NTHL1
  • Clonality:
    Recombinant
  • Clone:
    R08-8E9
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human NTHL1.
  • Pk:
    100 µl

Specifications

About this item