Anti-GRIN1 Rabbit Recombinant Antibody [clone: R08-4B6]
ABNORAB02057.100UL
: Abnova
- Antigen symbol:GRIN1
- Clonality:Recombinant
- Clone:R08-4B6
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human GRIN1.
- Pk:100 µl