Anti-PAK1 Rabbit Recombinant Antibody [clone: R08-5I5]
Catalog # ABNORAB02024.100UL
Supplier: Abnova
Specifications
- Antigen symbol:PAK1
- Clonality:Recombinant
- Clone:R08-5I5
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human PAK1.
- Pk:100 µl