Anti-AMPK alpha 2 Rabbit Recombinant Antibody [clone: R06-2E8]
Catalog # ABNORAB01608.100UL
Supplier: Abnova
Specifications
- Antigen symbol:AMPK alpha 2
- Clonality:Recombinant
- Clone:R06-2E8
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human AMPK alpha 2.
- Pk:100 µl