Anti-SKP2 Rabbit Recombinant Antibody [clone: R08-0A2]
ABNORAB01361.100UL
: Abnova
- Antigen symbol:SKP2
- Clonality:Recombinant
- Clone:R08-0A2
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human SKP2.
- Pk:100 µl