Anti-PNKP Rabbit Recombinant Antibody [clone: R03-0B8]
Catalog # ABNORAB01382.100UL
Supplier: Abnova
Specifications
- Antigen symbol:PNKP
- Clonality:Recombinant
- Clone:R03-0B8
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human PNKP.
- Pk:100 µl