Anti-FRS2 Rabbit Recombinant Antibody [clone: R01-7G7]
ABNORAB01450.100UL
: Abnova
- Antigen symbol:FRS2
- Clonality:Recombinant
- Clone:R01-7G7
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human FRS2.
- Pk:100 µl