Anti-NEK6 Rabbit Recombinant Antibody [clone: R01-4F7]
Catalog # ABNORAB01402.100UL
Supplier: Abnova
Specifications
- Antigen symbol:NEK6
- Clonality:Recombinant
- Clone:R01-4F7
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human NEK6.
- Pk:100 µl