Anti-ISG15 Rabbit Recombinant Antibody [clone: R08-1H5]
Catalog # ABNORAB02160.100UL
Supplier: Abnova
Specifications
- Antigen symbol:ISG15
- Clonality:Recombinant
- Clone:R08-1H5
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human ISG15.
- Pk:100 µl