Anti-IGF1R Rabbit Recombinant Antibody [clone: R06-6B1]
ABNORAB02171.100UL
: Abnova
- Antigen symbol:IGF1R
- Clonality:Recombinant
- Clone:R06-6B1
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human IGF1R.
- Pk:100 µl