Anti-UBE2D3 Rabbit Recombinant Antibody [clone: R01-4J4]
ABNORAB01846.100UL
: Abnova
- Antigen symbol:UBE2D3
- Clonality:Recombinant
- Clone:R01-4J4
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against human UBE2D3.
- Pk:100 µl