Order Entry
Poland
ContactUsLinkComponent
Anti-TRIM56 Rabbit Recombinant Antibody [clone: R02-8A3]
Anti-TRIM56 Rabbit Recombinant Antibody [clone: R02-8A3]
Catalog # ABNORAB01852.100UL
Supplier:  Abnova
Anti-TRIM56 Rabbit Recombinant Antibody [clone: R02-8A3]
Catalog # ABNORAB01852.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    TRIM56
  • Clonality:
    Recombinant
  • Clone:
    R02-8A3
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human TRIM56.
  • Pk:
    100 µl

Specifications

About this item