Anti-Collagen VI Rabbit Recombinant Antibody [clone: R09-8I1]
ABNORAB01496.100UL
: Abnova
- Antigen symbol:Collagen VI
- Clonality:Recombinant
- Clone:R09-8I1
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human Collagen VI.
- Pk:100 µl