Anti-beta Actin Rabbit Recombinant Antibody [clone: R09-4C9]
ABNORAB01561.100UL
: Abnova
- Antigen symbol:beta Actin
- Clonality:Recombinant
- Clone:R09-4C9
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human beta Actin.
- Pk:100 µl