Anti-Bcl-XL Rabbit Recombinant Antibody [clone: R02-2D3]
Catalog # ABNORAB01564.100UL
Supplier: Abnova
Specifications
- Antigen symbol:Bcl-XL
- Clonality:Recombinant
- Clone:R02-2D3
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human Bcl-XL.
- Pk:100 µl