Anti-CDK1/CDK2 Rabbit Recombinant Antibody [clone: R06-1A5]
ABNORAB01793.100UL
: Abnova
- Antigen symbol:CDK1/CDK2
- Clonality:Recombinant
- Clone:R06-1A5
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human CDK1/CDK2.
- Pk:100 µl