Anti-ENO1 Rabbit Recombinant Antibody [clone: R01-4C8]
Catalog # ABNORAB01271.100UL
Supplier: Abnova
Specifications
- Antigen symbol:ENO1
- Clonality:Recombinant
- Clone:R01-4C8
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against human ENO1.
- Pk:100 µl