Order Entry
Poland
ContactUsLinkComponent
Anti-PTEN Rabbit Recombinant Antibody [clone: R02-9D3]
Anti-PTEN Rabbit Recombinant Antibody [clone: R02-9D3]
Catalog # ABNORAB01220.100UL
Supplier:  Abnova
Anti-PTEN Rabbit Recombinant Antibody [clone: R02-9D3]
Catalog # ABNORAB01220.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    PTEN
  • Clonality:
    Recombinant
  • Clone:
    R02-9D3
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human PTEN.
  • Pk:
    100 µl

Specifications

About this item