Anti-PDPK1 (pSer241) Rabbit Monoclonal Antibody [clone: F7] (FITC)
Catalog # ABNOMAB23486.100
Supplier: Abnova
Specifications
- Antigen symbol:PDPK1
- Clonality:Monoclonal
- Clone:F7
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Reactivity:Human
- Modification:pSer241
- Immunogen:Rabbit monoclonal antibody raised against synthetic phosphopeptide of human PDPK1
- Pk:100 AU