Order Entry
Poland
ContactUsLinkComponent
AmberChrom™ chloride form 1X8 20-50 mesh, Supelco®
AmberChrom™ chloride form 1X8 20-50 mesh, Supelco®
Catalog # SIAL44324-100G
Supplier:  Merck
CAS Number:  
AmberChrom™ chloride form 1X8 20-50 mesh, Supelco®
Catalog # SIAL44324-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
CAS Number:69011-19-4
 :MFCD00132718
Formula:(C₁₀H₁₂·C₁₀H₁₀·C₈H₈·C₃H₉N)x