0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
AmberChrom™ Na+ form
AmberChrom™ Na+ form 50WX2 100-200 mesh, Supelco®
AmberChrom™ Na+ form 50WX2 100-200 mesh, Supelco®
AmberChrom™ Na+ form 50WX2 100-200 mesh, Supelco®
Catalog # SIAL44462-100G
Supplier:
Merck
CAS Number:
Expand image
AmberChrom™ Na+ form 50WX2 100-200 mesh, Supelco®
Catalog # SIAL44462-100G
Supplier:
Merck
CAS Number:
Specifications
Pk:
100 g
Seq:
3401
Specifications
About this item
Product Details
:
CAS Number:
69011-22-9
Product Details