Polyvinyl alcohol USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:9002-89-5
:25213-24-5
Formula:[-CH₂CH(OH)-]n
MDL Number:MFCD00081922
Storage Temperature:Ambient
Specification Test Results
- Viscosity Method Used:As Reported
- pH of a 4% Solution:5.0 - 8.0
- Degree of Hydrolysis:85 - 89%
- Loss On Drying:max. 5.0%
Specification Test Results
Frequently Bought Together