Order Entry
Poland
ContactUsLinkComponent
 
(±)-2-Methyl-2,4-pentanediol ≥98%, Reagent Grade
  SPCMHE188-20LT
 :  
(±)-2-Methyl-2,4-pentanediol ≥98%, Reagent Grade
  SPCMHE188-20LT
 :  

 

  • Pack type:
    Steel can
  • Pk:
    20 L
  • Seq:
    5302

 

 

  • Assay:
    98% Min.

 

 

 :
 :118.18 g/mol
 :13,04730
 :198 °C (1013 hPa)
 :–40 °C
 :0,923 g/cm³ (20 °C)
 :93 °C
 :Ambient
 :203-489-0
 :GHS07
 :107-41-5
 :C₆H₁₄O₂
 :MFCD00004547