Order Entry
Poland
ContactUsLinkComponent
DL-Malic acid ≥99.0% FCC
Irritant
DL-Malic acid ≥99.0% FCC
Catalog # SPCMMA147-500GM
Supplier:  Spectrum Chemical
CAS Number:  
undefined
Irritant
DL-Malic acid ≥99.0% FCC
Catalog # SPCMMA147-500GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (C₄H₆O₅):
    99.0 - 100.5%
  • Optical Rotation:
    -0.10 to +0.10
  • Fumaric Acid:
    max. 1.0 %
  • Lead (Pb):
    max. 2 mg/kg
  • Maleic Acid:
    max. 0.05 %

Specification Test Results

About this item

 :
 :134.09 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:617-48-1
Formula:HOOCCH(OH)CH₂COOH
MDL Number:MFCD00064212