Order Entry
Poland
ContactUsLinkComponent
Zinc sulphate monohydrate ≥98% FCC
Corrosive
Irritant
Environment
Zinc sulphate monohydrate ≥98% FCC
Supplier:  Spectrum Chemical
Certificates
Corrosive
Irritant
Environment
Zinc sulphate monohydrate ≥98% FCC
Supplier:  Spectrum Chemical

Specification Test Results

  • Assay:
    98.0 - 100.5%
  • ACIDITY:
    TO PASS TEST
  • ALKALIES AND ALKALINE EARTHS:
    max. 0.5 %
  • CADMIUM (Cd):
    max. 5 mg/kg
  • LEAD (Pb):
    max. 4 mg/kg
  • MERCURY:
    max. 5 mg/kg
  • SELENIUM (Se):
    max. 0.003 %
  • IDENTIFICATION:
    TO PASS TEST

Specification Test Results

Frequently Bought Together