Anti-MHC Class 2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-8481R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:MHC Class 2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:111-200/263
- Form:Liquid
- Gene ID:3109
- Antigen synonyms:MHC class II antigen DMB|HLA-DMB|D6S221E|Really interesting new gene 7 protein|HLA class II histocompatibility antigen, DM beta chain|DMB|RING7
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:27 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human HLA class II histocompatibility antigen DM beta chain
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Plays a critical role in catalyzing the release of class II-associated invariant chain peptide (CLIP) from newly synthesised MHC class II molecules and freeing the peptide binding site for acquisition of antigenic peptides. In B-cells, the interaction between HLA-DM and MHC class II molecules is regulated by HLA-DO.
Type: Primary
Antigen: MHC Class 2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 111-200/263
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse