Anti-EDA2R Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-7111R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:EDA2R
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Environmentally Preferable:
- Epitope:51-150/297
- Form:Liquid
- Gene ID:60401
- Antigen synonyms:X-linked ectodysplasin-A2 receptor|UNQ2448/PRO5727/PRO34080|XEDAR|Tumor necrosis factor receptor superfamily member 27|EDA2R|EDA-A2 receptor|EDA-A2R|TNFRSF27|EDAA2R
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:33 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human EDA2R/TNFRSF27
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Receptor for EDA isoform A2, but not for EDA isoform A1. Mediates the activation of the NF-kappa-B and JNK pathways. Activation seems to be mediated by binding to TRAF3 and TRAF6.
Type: Primary
Antigen: EDA2R
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/297
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse