Specifications
- Antibody type:Primary
- Antigen name:SLC38A2/SNAT2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:21-150/506
- Form:Liquid
- Antigen synonyms:PRO1068|Slc38a2|System A amino acid transporter 2|KIAA1382|Sodium-coupled neutral amino acid transporter 2|ATA2|Amino acid transporter A2|SNAT2|System A amino acid transporter|System A transporter 1|Solute carrier family 38 member 2|System N amino acid transporter 2|S38A2_HUMAN|SAT2|Amino acid transporter 2|Protein 40-9-1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:56 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human SLC38A2/SNAT2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The sodium-coupled neutral amino acid transporters (SNAT) of the SLC38 gene family include System A subtypes SNAT1, SNAT2 and SNAT4 and System N subtypes SNAT3 and SNAT5. The SLC38 transporters are essential for the uptake of nutrients, energy production, metabolism, detoxification, and the cycling of neurotransmitters. SNAT2, also designated ATA2, PRO1068 and SAT2 is encoded by the human gene SLC38A2. The functional role of SNAT2 in the nervous system is unclear. Protein expression is notably enriched in the spinal cord and brain stem nuclei of the auditory system. System A transport proteins are also present in placental tissue. These SNAT proteins may play a significant role in fetal development and inhibition of the transport system has been associated with fetal growth retardation.
Type: Primary
Antigen: SLC38A2/SNAT2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-150/506
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat