Anti-Glycoprotein VI Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-9897R-A680
: Bioss
- Antibody type:Primary
- Antigen name:Glycoprotein VI
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:121-220/339
- Form:Liquid
- Gene ID:51206
- Antigen synonyms:GP6|GPIV|MGC138168|GPVI_HUMAN|Platelet collagen receptor|Glycoprotein VI|Platelet glycoprotein VI precursor|Platelet glycoprotein VI|GPVI
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GPVI/Glycoprotein VI
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Glycoprotein VI (GPVI) is a membrane glycoprotein receptor for collagen found in platelets. GPVI plays an important role in platelet procoagulant activity which may contribute to arterial and venous thrombus formation. GPVI is expressed in megakaryocytes and platelets. Patients deficient in GPVI suffer from mild hemorrhagic diathesis and their platelets fail to aggregate in response to collagen. There are three isoforms.
Type: Primary
Antigen: Glycoprotein VI
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 121-220/339
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat