Anti-Mitoferrin 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-9523R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Mitoferrin 1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:201-300/338
- Form:Liquid
- Gene ID:51312
- Antigen synonyms:Mitochondrial solute carrier protein|predicted protein of HQ2217|MSCP|PRO1584|PRO2217|HT015|Mitoferrin-1|SLC25A37|Mitochondrial iron transporter 1|Solute carrier family 25 member 37|PRO1278|MSC|Mitoferrin1|MFRN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Mitoferrin 1
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Mitoferrin1, SLC25A37, belongs to the mitochondrial carrier family and contains 3 Solcar repeats. It is a mitochondrial iron transporter that specifically mediates iron uptake in developing erythroid cells and plays an essential role in heme biosynthesis.
Type: Primary
Antigen: Mitoferrin 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/338
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat