Anti-Ceramide synthase 2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-5077R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Ceramide synthase 2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:201-300/380
- Form:Liquid
- Gene ID:29956
- Antigen synonyms:Ceramide synthase 2|Tumor metastasis-suppressor gene 1 protein|SP260|CerS2|LAG1 longevity assurance homolog 2|L3|TMSG1|LASS2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:45 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Ceramide synthase 2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
LASS2 (LAG1 homolog, ceramide synthase 2) has sequence similarity to yeast longevity assurance gene 1. It is thought to be involved in sphingolipid synthesis. Expression of LASS2 is transiently increased during the period of active myelination and is specifically localised to white matter tracts of the brain, including the Schwann cells of sciatic nerves suggesting that LASS2 is important for the synthesis of dihydroceramide used for synthesis of myelin sphingolipids. Expression of LASS2 in hepatocellular carcinoma cell lines suppresses the growth of cancer cells.
Type: Primary
Antigen: Ceramide synthase 2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/380
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat