Anti-TREM2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-2723R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:TREM2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Pig,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:1-100/230
- Form:Liquid
- Gene ID:54209
- Antigen synonyms:Trem2b|TREM2|TREM-2|Trem2a|Triggering receptor expressed on monocytes 2|Triggering receptor expressed on myeloid cells 2|Trem2c
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:25 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human TREM2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
May have a role in chronic inflammations and may stimulate production of constitutive rather than inflammatory chemokines and cytokines. Forms a receptor signaling complex with TYROBP and triggers activation of the immune responses in macrophages and dendritic cells.
Type: Primary
Antigen: TREM2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/230
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat; Pig