Anti-IGIP Rabbit Polyclonal Antibody (ALEXA FLUOR® 647)
BOSSBS-15574R-A647
: Bioss
- Antibody type:Primary
- Antigen name:IGIP
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Epitope:21-53/53
- Form:Liquid
- Antigen synonyms:C5orf53|IgA-inducing protein homolog (Bos taurus)|LOC492311|IGIP|IGIP_HUMAN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:6 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human IGIP
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
IGIP.
Type: Primary
Antigen: IGIP
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 647
Public Immunogen Range: 21-53/53
Host: Rabbit
Isotype: IgG
Reactivity: