Anti-IGF2AS Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)
Catalog # BOSSBS-15568R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:IGF2AS
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:101-168/168
- Form:Liquid
- Antigen synonyms:IGF2 Antisense RNA|IGF2AS|PEG8/IGF2AS Protein|IGF2 Antisense RNA Non-Protein Coding|PEG8|IGF2 Antisense Gene Protein 1|Putative Insulin-Like Growth Factor 2 Antisense Gene Protein 1|IGF2 AS1|Insulin Like Growth Factor 2, Antisense|IGF2 Antisense RNA 1|IG2AS_HUMAN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:18 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human IGF2AS
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This gene expresses a paternally imprinted antisense transcript of the insulin-like growth factor 2 gene. The transcript is overexpressed in Wilms' tumor. This gene is predicted to be non-coding because the predicted protein is not conserved in any other species and the majority of transcripts would be candidates for non-sense mediated decay (NMD) if a protein were expressed. Alternative splicing results in multiple transcript variants.
Type: Primary
Antigen: IGF2AS
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 101-168/168
Host: Rabbit
Isotype: IgG
Reactivity: