Anti-GPRASP2 Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)
Catalog # BOSSBS-15393R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:GPRASP2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:201-300/838
- Form:Liquid
- Antigen synonyms:GASP2_HUMAN|FLJ37327|G-protein coupled receptor-associated sorting protein 2|GASP2|FLJ35662|GPRASP2|GASP-2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:94 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GPRASP2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
GPRASP2.
Type: Primary
Antigen: GPRASP2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 201-300/838
Host: Rabbit
Isotype: IgG
Reactivity: