Anti-GPR89A Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)
Catalog # BOSSBS-15391R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:GPR89A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Environmentally Preferable:
- Epitope:221-320/445
- Form:Liquid
- Gene ID:51463
- Antigen synonyms:GPHR|Protein GPR89B|GPR89|Putative NF-kappa-B-activating protein 90|HSPC201|Protein GPR89A|GPHRB|GPHRA_HUMAN|Golgi pH regulator A|GPHRA|Putative MAPK-activating protein PM01|UNQ192/PRO218|Golgi pH regulator B|CGI-13|SH120|GPR89B|GPR89A
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:53 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GPR89A
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
GPR89A is a nearly identical copy of the GPR89B gene (MIM612806).
Type: Primary
Antigen: GPR89A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 221-320/445
Host: Rabbit
Isotype: IgG
Reactivity: Rat