Order Entry
Poland
ContactUsLinkComponent
Anti-C6ORF136 Rabbit Polyclonal Antibody (FITC)
Anti-C6ORF136 Rabbit Polyclonal Antibody (FITC)
Catalog # BOSSBS-15219R-FITC
Supplier:  Bioss
Anti-C6ORF136 Rabbit Polyclonal Antibody (FITC)
Catalog # BOSSBS-15219R-FITC
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C6orf136
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Epitope:
    131-230/315
  • Form:
    Liquid
  • Gene ID:
    221545
  • Antigen synonyms:
    Uncharacterized protein C6orf136|Chromosome 6 open reading frame 136|CF136_HUMAN
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    36 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human C6ORF136
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Making up nearly 6% of the human genome, chromosome 6 contains around 1200 genes within 170 million base pairs of sequence. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer suggesting the presence of a cancer susceptibility locus. Porphyria cutanea tarda is associated with chromosome 6 through the HFE gene which, when mutated, predisposes an individual to developing this porphyria. Notably, the PARK2 gene, which is associated with Parkinson's disease, and the genes encoding the major histocompatiblity complex proteins, which are key molecular components of the immune system and determine predisposition to rheumatic diseases, are also located on chromosome 6. Stickler syndrome, 21-hydroxylase deficiency and maple syrup urine disease are also associated with genes on chromosome 6. A bipolar disorder susceptibility locus has been identified on the q arm of chromosome 6. The C6orf136 gene product has been provisionally designated C6orf136 pending further characterisation.

Type: Primary
Antigen: C6orf136
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 131-230/315
Host: Rabbit
Isotype: IgG
Reactivity: Mouse