Order Entry
Poland
ContactUsLinkComponent
Leukotriene C₄ ELISA Kit
Leukotriene C₄ ELISA Kit
Supplier:  Cayman Chemical
Certificates
undefined
Leukotriene C₄ ELISA Kit
Supplier:  Cayman Chemical

Specifications

  • Assay Type:
    Competitive
  • Mid point:
    175 - 275 pg/ml
  • Detection range:
    17,1 - 5000 pg/ml
  • Sensitivity:
    40 pg/ml

Specifications

Related Information

Frequently Bought Together