Order Entry
Poland
ContactUsLinkComponent
 
Aspartate Aminotransferase (AST) (Human) ELISA Kit
  ABCAAB285299-96
 :  Abcam
 :  
undefined
Aspartate Aminotransferase (AST) (Human) ELISA Kit
  ABCAAB285299-96
 :  Abcam
 :  

 

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Description:
    Aspartate Aminotransferase (AST) (Human) ELISA kit
  • Tests per kit:
    100 assays

 

 

A Sandwich ELISA kit to the quantify of human AST in serum and other biological fluids.

  • Easy, convenient and time-saving method to measure the level of AST in human serum, plasma, tissue homogenates, culture supernatants and other biological fluids.
  • Detection Range: 0,313 - 20 ng/ml
  • Sensitivity: < 0,188 ng/ml
  • Assay Precision: Intra-Assay: CV < 8%; Inter-Assay: CV < 10%

Biosynthesis of L-glutamate from L-aspartate or L-cysteine. Important regulator of levels of glutamate, the major excitatory neurotransmitter of the vertebrate central nervous system. Acts as a scavenger of glutamate in brain neuroprotection. The aspartate aminotransferase activity is involved in hepatic glucose synthesis during development and in adipocyte glyceroneogenesis. Using L-cysteine as substrate, regulates levels of mercaptopyruvate, an important source of hydrogen sulfide. Mercaptopyruvate is converted into H2S via the action of 3-mercaptopyruvate sulfurtransferase (3MST). Hydrogen sulfide is an important synaptic modulator and neuroprotectant in the brain. BioVision’s AST ELISA kit is a sandwich ELISA assay for the quantitative measurement of human AST in serum, plasma and cell culture supernatants. The density of color is proportional to the amount of human AST captured from the samples.