

Potassium permanganate concentrate for dilution to 0.02 mol (0.1N) in aqueous solution, ConvoL NORMADOSE® volumetric solution
numer katalogowy 91352.600
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass ampoule
- Pk:1 Ampoul
- Seq:0501
Specifications
Specification Test Results
- Molarity:0.0199 - 0.0201 mol/l (20°C)
Specification Test Results
About this item
:
:158.03 g/mol
:Dose for preparation of 1 litre of solution.
:GHS05
:GHS09
Storage Temperature:Ambient
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364