

Petroleum spirit, 30…50 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849
Boiling Pt:30…50 °C (1013 hPa)
Density:0.65 g/cm3 (20 °C)
Flash Pt:<–30 °C
UN:1268
ADR:3,I
:GHS02
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Acidity:max. 0.0002 meq/g
- Boiling range:30 - 50 °C
- Iodine value:max. 0.3
- Aromatics (as C6H6):max. 100 ppm
- Evaporation residue:max. 30 ppm
- Sulphur compounds (as S):max. 50 ppm
- Water :max. 100 ppm
- Al (Aluminium):max. 0.5 ppm
- B (Boron):max. 0.02 ppm
- Ba (Barium):max. 0.1 ppm
- Ca (Calcium):max. 0.5 ppm
- Cd (Cadmium):max. 0.05 ppm
- Co (Cobalt):max. 0.02 ppm
- Cr (Chromium):max. 0.02 ppm
- Cu (Copper):max. 0.02 ppm
- Fe (Iron):max. 0.1 ppm
- Mg (Magnesium):max. 0.1 ppm
- Mn (Manganese):max. 0.02 ppm
- Ni (Nickel):max. 0.02 ppm
- Pb (Lead):max. 0.1 ppm
- Sn (Tin):max. 0.1 ppm
- Zn (Zinc):max. 0.1 ppm
Specification Test Results
Related Information
Frequently Bought Together