2’-Bromo-5’-(trifluoromethoxy)-2,2,2-trifluoroacetophenone ≥95%
Supplier: Apollo Scientific
Certificates
About this item
:
Formula:C₉H₃BrF₆O₂
:337.013 g/mol
Frequently Bought Together
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Methanol ≥99.5% (by GC), EMPROVE® ESSENTIAL NF, Ph. Eur., BP, JPE, extra pure suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
BENZENE FOR GC 1 * 1 L
Supplier: CHEMPUR
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dibutylamine for HPLC
Supplier: Thermo Fisher Scientific
Ethanol for histology, J.T.Baker®
Supplier: Avantor
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Carbon disulphide 99.9% for HPLC
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide 99.8+%, extra pure
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% for HPLC
Supplier: VWR Chemicals
n-Hexane for HPLC
Supplier: POCH AVANTOR BRAND
Chlorobenzene 99+%, pure
Supplier: Thermo Fisher Scientific
Chloroform 99.8% stabilised for analysis
Supplier: Thermo Fisher Scientific