2-Bromo-6-fluoro-3-iodobenzoic acid ≥95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2090463-67-3
:
Formula:C₇H₃BrFIO₂
:344.90439 g/mol
Frequently Bought Together
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
N-Ethyldiisopropylamine, Sigma-Aldrich®
Supplier: Merck
Xylene (mixture of isomers) for histology
Supplier: Honeywell Chemicals
Xylene (mixture of isomers), pure analytical reagent
Supplier: CHEMPUR
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
1,3-Dimethyl-2-imidazolidinone ≥99.5% (by GC), CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane 99.9% stabilised, AcroSeal® for biochemistry
Supplier: Thermo Fisher Scientific
Aeroflow tap, na, general spare part 1 * 1 items
Supplier: Avantor