Methyl 5-bromo-4-methoxy-2-methylbenzoate ≥95%
:1131587-94-4
:
:C₁₀H₁₁BrO₃
:MFCD11112987
:259.1 g/mol
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Glycerine 85% Ph. Eur.
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals