Metal-In Shielding Bubble Bags
: DESCO
- Combines 'Faraday Cage' and mechanical protection
- Shields about twice as well as normal shielding bag of equivalent size
- Flap can be tucked in to enclose contents or folded down and held in place by a label
- Inner and outer surface resistance: 1×104 to <1×1011 ohms; T₁₀₀₀: <2 sec at 50% rH
Frequently Bought Together