Union Fittings, Hose Barb, Straight, Polypropylene, Dynalon®
Supplier: Avantor Fluid Handling
Certificates
About this item
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Chemical resistant
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX06288-20BLK
- MFLX30601-00BLK
- Orifice dimensions:
- 3 mm (0.125")
- 2 mm (0.078125")
- Sterilisation:
- Autoclave only
- Autoclave only
- Warranty:
- 1 year
- Out of box